Learn More
Koettermann™ Exploris™ Mobile Cabinet
Product Code. 15508505
Change view
Click to view available options
:
Mobile Cabinet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15508505 Mobile Cabinet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15508505 Supplier Koettermann™ Supplier No. 31500106
Currently Unavailable: We have no information when this product will be available.
View alternative products
Includes
Four drawers
Offers adaptability for wide range of applications.
  • Underbench cabinets
  • Lockable casters
  • Equipped with different front panels
Specifications
Color White
Depth (Metric) 51.6 cm
Dimensions (D x W x H) 51.6 x 120 x 74 cm
Height (Metric) 74 cm
Width (Metric) 120 cm
Material Steel