Learn More
Koettermann™ Exploris™ Balance Bench
Product Code. 14855421
Change view
Click to view available options
Balance Bench
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
14855421 Balance Bench
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 14855421 Supplier Koettermann™ Supplier No. 50900001
  • ONLINE EXCLUSIVE PRICE
    2640.00€
    2146.09€ / Pack of 1
    Save 493.91€
    18% Off
View all promotions
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products
Designed to attenuate external vibrations.
  • Stand-alone balance
  • Attenuate external vibrations
  • Large mass, oscillation-damped mounting
  • Low-vibration balance platform
  • Balance block mechanically separated from the outer chassis of balance bench
  • Vibrations caused by impact are not transferred directly to balance block
Specifications
Dimensions (L x W x H) 60 x 90 x 75 cm
Type Balance Bench