missing translation for 'onlineSavingsMsg'
Learn More

CELLPATH EXPERT XTF MOUNTING MEDIUM

Product Code. p-200113593
Click to view available options
Volume (Metric):
1 L
4 x 250 mL
Unit Size:
1 L
Pack of 4
This item is not returnable. View return policy

Product Code. 30226294

Brand: CELLPATH SEA190000A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ExPert XTF is a xylene and toluene free mounting medium which is used to coverslip and permanently preserve thin stained tissue sections. Being xylene and toluene free (both of which can be carcinogenic), XTF provides enhanced safety to the end-user.

  • The refractive index is similar to ExPert being close to that of glass at +20°C.
  • Key Benefits:
  • Safe for end user as xylene and toluene free
  • Suitable for manual and automated cover slipping

Specifications

Product Type Mounting Medium
Reagent Type Mounting Solution
Volume (Metric) 1 L
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • ExPert XTF
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P370+P378-In case of fire: Use sand, carbon dioxideor powder extinguisher to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contains : XTF Clearing Agent,Isopar E, Isobutyl methacrylate, 2-ethylhexyl methacrylate,1-dodecanethiol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.