Learn More
BINDER™ Expansion Rack
Product Code. 15631636 Shop All BINDER Products
Click to view available options
Material:
Aluminum
For Use With (Equipment):
VD 53, VDL 53
Unit Size:
Each
Product Code. 15631636
Brand: BINDER™ 80090371
This item requires a price update prior to being ordered.
Request a Price for this item here.
Material: Aluminum; For Use With: VD 53, VDL 53
  • Constructed of aluminum or stainless steel
  • Expansion rack for vacuum drying chambers
Specifications
Description Expansion rack - aluminum
For Use With (Equipment) VD 53, VDL 53
Material Aluminum
Type Rack