Learn More
Lenz Laborglasintrumente Expansion Adapters with Ground Joints
Click to view available options
Cone Size:
NS 14/23
Unit Size:
Each
Description
- Acc. to DIN 12257
- Conical ground sockets and cones
- Made of DURAN™ tubing
Specifications
Specifications
| Cone Size | NS 14/23 |
| Material | Duran Glass |
| Standard Taper Joint No. | Socket NS 24/29, Cone NS 14/23 |