Learn More
Applied Biosystems™ ExoSAP-IT™ Express PCR Product Cleanup Reagent
Product Code. p-7092196
Click to view available options
No. of Reactions:
100
2000
480 reactions, 8-tube strip
500
5000
Unit Size:
100 reactions
2000 reactions
480 reactions
500 reactions
5000 reactions
Product Code. 15543677
Brand: Applied Biosystems™ 75001.1.ML
Removes excess primers and dNTPs. Does not interfere with downstream applications.
  • Mixture of two hydrolytic enzymes, a modified Exonuclease I and Shrimp Alkaline Phosphatase (SAP), in a specially formulated buffer
  • One-tube, one-step PCR cleanup
  • Five-minute protocol
  • Single tube, 8-tube strip, and 96-well plate formats offer flexibility from manual pipetting to automated workflows
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C
No. of Reactions 500