Learn More
Excelta™ Precision Tweezers with Straight Strong Medium Tips
Precision Tweezers feature strong tips for long life.
23.03€
Specifications
Product Type Tweezers, Electronic
Quantity (No. per Case) 1 Ea.
Height (English) Body 0.015 in.
Height (Metric) Body 0.03 cm
Width (English) Body 0.32 in.
View More Specs
Products 1
Product Code Brand Design Length (Metric) Material Precision Price Quantity & Availability  
Product Code Brand Design Length (Metric) Material Precision Price Quantity & Availability  
11723266
View Documents Promotion Details
Excelta™
00D
00D 120 mm Carbon Steel High
23.03€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
Description
Strong tips for long life
  • Anti-magnetic stainless steel or Carbon Steel
Specifications
Specifications
Tweezers, Electronic
0.015 in.
0.32 in.
1 Ea.
0.03 cm
0.8 cm