Learn More
Excelta™ Precision Tweezers with Straight Strong Medium Tips
Product Code. 11723266
Encompass_Preferred
Click to view available options
Length (Metric):
120 mm
Material:
Carbon Steel
Precision:
High
Design:
00D
Unit Size:
Each
Product Code. 11723266
Brand: Excelta™ 00D
Only null left
Add to basket
Add to basket
Precision Tweezers feature strong tips for long life.
Strong tips for long life
  • Anti-magnetic stainless steel or Carbon Steel
Specifications
Tip Style Straight Strong, Serrated
Design 00D
Length (English) 4.75 in.
Material Carbon Steel
Length (Metric) 120 mm
Product Type Tweezers, Electronic
Quantity (No. per Case) 1 Ea.
Height (English) Body 0.015 in.
Height (Metric) Body 0.03 cm
Precision High
Thickness 0.09 in.
Width (English) Body 0.32 in.
Width (English) Tip 0.0177 in.
Width (Metric) Body 0.8 cm
Width (Metric) Tip 0.044 cm
Thickness (English) Tip 0.0098 in.
Thickness (Metric) Tip 0.249 mm
Style High-precision
Show More Show Less
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.