Learn More
R&D Systems™ ExCellerate iPSC Expansion Medium
Product Code. 18759296
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18759296 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18759296 Supplier R&D Systems™ Supplier No. CCM036
  • ONLINE EXCLUSIVE PRICE
    307.00€
    289.80€ / 500mL
    Save 17.20€
    5% Off
 Restricted Item
Estimated Shipment: 09-11-2026
Specifications
Product Line R&D Systems
Quantity 500 mL
Regulatory Status Animal Free
Species Human
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.