Learn More
R&D Systems™ ExCellerate Human T Cell Expansion Media, Xeno-Free
Product Code. 18777146
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18777146 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18777146 Supplier R&D Systems™ Supplier No. CCM030
  • ONLINE EXCLUSIVE PRICE
    146.59€
    130.20€ / 500mL
    Save 16.39€
    11% Off
 Restricted Item
Estimated Shipment: 16-12-2026
Media for the ex vivo expansion and culture of human T cell lymphocytes under xeno-free conditions
  • Robust T cell expansion under serum-free and xeno-free conditions
  • Optimized for use with the Cloudz™ T Cell Activation Kit
  • Compatible with a variety of T cell activation methods
  • Supports T cell expansion from PBMCs or purified CD3+ T cells
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) T-Cell Expansion
Product Line R&D Systems
Quantity 500 mL
Content And Storage Store the unopened product at 2 - 8°C. Do not use past expiration date.
Description Facilitates discovery and preclinical research for T cell immunoregulation and therapy, robust platforms for ex vivo expansion and maintenance of T cells, including specialized serum-free or xeno-free media