Learn More
R&D Systems™ ExCellerate B Cell Media, Xeno-Free
Product Code. 18776036
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18776036 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18776036 Supplier R&D Systems™ Supplier No. CCM031
  • ONLINE EXCLUSIVE PRICE
    159.83€
    141.75€ / 500mL
    Save 18.08€
    11% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Expansion and clonal growth of B cells.
TRUSTED_SUSTAINABILITY
Specifications
Product Line R&D Systems
Quantity 500 mL
Content And Storage Store the unopened product at 2 to 8 °C. Do not use past expiration date
Product Title
Select an issue
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.