Learn More
Cytiva ExcelGel SDS Buffer Strips
Product Code. 11515015 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
6 Pack
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11515015 6 Pack Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11515015 Supplier Cytiva Supplier No. 17134201
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Semi dry precast buffer strips for horizontal SDS electrophoresis. Cytiva Lifescience™ ExcelGel SDS Buffer Strips are designed for use with ExcelGel Precast Gels.
  • Designed for use with ExcelGel Precast Gels for SDS-PAGE
  • Include strips for both anode and cathode
TRUSTED_SUSTAINABILITY
Specifications
Formulation Acrylamide 0.55%
For Use With (Application) SDS-PAGE
Quantity 6 Pack
Storage Requirements Cool, dry place
Product Type ExcelGel SDS Buffer strips
For Use With (Equipment) Multiphor II Flatbed Electrophoresis System
compliance-icons
Product Identifier
  • ExcelGel SDS Buffer Strips
Signal Word
  • Danger
Hazard Category
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
Hazard Statement
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • EUH208-Contains Acrylamide. May produce an allergic reaction.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Acrylamide
  • UNKNOWN ACUTE-75 per cent of the mixture consists of component(s) of unknown acute toxicity.