Learn More
Klever Excel Plus Metal Detectable
Click to view available options
Color:
Blue
Unit Size:
Pack of 50
Description
- Due to the wider blade feeder, even thicker material can be cut easily and safely.
- Due to the metal-detectable plastic, the safety knife is very well suited for use in the food, chemical or pharmaceutical industry.
- The disposable product cuts up to double-wall corrugated cardboard, film, adhesive tape, strapping tape, etc.
- With practical eyelet for lanyard fastening or hanging.
Specifications
Specifications
| Product Type | Safety Knife |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Color | Blue |
Product Content Correction
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur