missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Evolution™ One/One Plus UV-Vis Spectrophotometers

Product Code. 17847047 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Evolution One INTL Security Bundle
Evolution One INTL USP/EP Standards Bundle
Evolution One Plus UV-Vis Spec
Evolution One US USP/EP Standards Bundle
Evolution One UV-Vis Spec
Evolution OnePlus INTL Security Bundle
Evolution OnePlus INTL USP/EP Standards Bundle
Spectral Bandwidth:
1 nm
1.0, 2.0 nm Variable
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Spectral Bandwidth Unit Size
17847047 Evolution One US USP/EP Standards Bundle 1 nm Each
17329263 Evolution One UV-Vis Spec 1 nm Each
17339263 Evolution One Plus UV-Vis Spec 1.0, 2.0 nm Variable Each
17435603 Evolution One INTL Security Bundle 1 nm Each
17445603 Evolution One INTL USP/EP Standards Bundle 1 nm Each
17465603 Evolution OnePlus INTL Security Bundle 1.0, 2.0 nm Variable Each
17455603 Evolution OnePlus INTL USP/EP Standards Bundle 1.0, 2.0 nm Variable Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 17847047 Supplier Thermo Scientific™ Supplier No. 840370200
  • ONLINE EXCLUSIVE PRICE
    30985.00€
    28312.36€ / Pack of 1
    Save 2672.64€
    8% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ Evolution™ UV-Vis Spectrophotometers combine reliable and versatile hardware with easy-to-learn, easy-to-use Thermo Scientific™ Insight™ Pro Software. Together they will meet the diverse needs your organization requires from an analytical tool.

For routine, complex research and advanced applications, including current compliance, the Thermo Scientific™ Evolution™ One/One Plus UV-Vis Spectrophotometer series brings together reliable, versatile hardware, a broad range of accessories, with easy-to-use, easy-to-learn Thermo Scientific™ Insight™ Pro Software.

Webinar: Identification of protein aggregation through UV-Vis spectroscopy

For routine to research analysis, this instrument series offers real innovations that provide enhanced usability and performance - without added complexity. With straightforward software, cutting-edge instrumentation and a wide selection of accessories, the Evolution One and One Plus UV-Vis Spectrophometers work with you to perform experiments the way you want - for the results you need.

Optimized Engineering Delivers Ultimate Performance:
  • Double-beam geometry with fixed 1nm or variable spectral bandwidth options
  • Application Focused Beam Geometry (AFBG) technology enables the best performance from microcells, solid samples, and fiber optic probes (Evolution One Plus Only)
  • Xenon flash lamp requires no warm-up time allowing instant measurements
  • Meet chemical, materials science, or biological sampling and measurement needs with a broad range of accessories
Insight Pro Software—Easy-to-learn, easy-to-use
  • Reduce training time: Optimized user interface with prompting software defines clear step-by-step workflow and is easy to teach and learn
  • Prevent costly sampling and measurement errors: Create dedicated workflows for complex assay and transform multi-step workflows into one simplified, easy-to-follow method suitable for any level of user with Customized User Environment (CUE)
  • Always current: Whether it is user inputs or application requirements, or new regulations, Insight Pro Software is continuously improving with quick upgrades to deliver greater capabilities and efficiencies for your lab’s needs
Accessories for all your Sampling Needs:
  • Wide accessory selection with various temperature control options, sample changers, sippers and solid sampling devices complete your analysis
  • Choice of Peltier Single Cell Holder or a Smart 8-Cell Peltier system for temperature control and sample monitoring from 0° to 100 °C
  • Easy software integration gives you complete control of your measurements

Regulatory/Quality Control Environments:
To use the EvolutionOne/One Plus UV-Vis Spectrophotometers in a regulated or quality control environment, we offer a wide selection of compliance products and services to reduce implementation time and assure system performance.

Evolution One/One Plus Security and Validator Bundles include:
  • Evolution One or One Plus UV-Vis Spectrometer
  • INSIGHT Pro with Security Software
  • Validator for Insight Pro US Ltr or INTL A4 Paper Version
  • Qualification Filter Kit for UV-Vis
Evolution One/One Plus USP/EP Standards Pharma Bundles include:
  • Evolution One or One Plus Spectrometer
  • INSIGHT Pro with Security Software
  • Validator for Insight Pro US Ltr or INTL A4 Paper Version
  • USP and EP UV Standards Set
  • USP and EP Visible Standards Set
TRUSTED_SUSTAINABILITY

Specifications

Product Line Evolution One
Spectral Bandwidth 1 nm
Type Evolution One US USP/EP Standards Bundle
Includes Evolution One Computer Control UV-Vis, INSIGHT Pro with Security 1.0, Validator for INSIGHT Pro 1.0 US Letter Paper Version, USP and EP UV Standards Set
- USP and EP Visible Standards Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.