Learn More
Thermo Scientific™ eVol™ XR Dispensing System Syringes
Product Code. p-4525955 Shop All Thermo Scientific Products
Click to view available options
Volume (Metric):
5 μL
50 μL
100 μL
500 μL
Unit Size:
Each
Product Code. 10558995
Brand: Thermo Scientific™ 66002021
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Rely on these versatile and functional syringes in sizes from 5 to 1000μL for the Thermo Scientific™ eVol™ Dispensing System.
Rely on exceptional versatility and functionality when using Thermo Scientific™ eVol™ XCHANGE Syringes. For use with the unique, hand-held eVol XR Dispensing System, the syringes are available in sizes from 5 to 1000μL. Integrated XCHANGE™ coupling allows you to change syringes quickly and easily. The addition of MEPS to XCHANGE syringe offers automated sample preparation by SPE in a handheld system. The eVol XR Dispensing System accurately and reproducibly performs a wide variety of liquid handling procedures
The eVol XR Dispensing System is a unique, hand-held digital dispenser that accurately and reproducibly performs a wide variety of liquid handling procedures. The easy-to-use device couples an analytical syringe to a programmable electronic controller, enabling highly reproducible dispensing for all users.
The eVol system is ideal for laboratories that need to repeatedly dispense nonaqueous liquids, offering greater accuracy and reproducibly for applications such as:
  • Routine dispensing of volatile solvents, hazardous, corrosive or viscous chemicals
  • Preparation of calibration standards
  • Addition of internal standards
The system unifies two precision devices: a digitally controlled electronic drive, and a Thermo Scientific™ XCHANGE™ enabled analytical syringe.
Repeatable Performance for Higher Confidence in Your Results
  • Can be calibrated easily by a user to ensure validity of results
  • Intuitive, easy-to-use interface with touch wheel and full-color screen
  • Complies with GLP and GMP guidelines
TRUSTED_SUSTAINABILITY
Specifications
Description eVol syringe, 5µL
Product Type Dispensing System Syringe
Volume (Metric) 5 μL
Product Line HyperSep
For Use With (Equipment) Electronic Dispensing System