Learn More
EVL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553864
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SSQRQVQNGP SPDEMDIQRR QVMEQHQQQR QESLERRTSA TGPILPPGHP SSAASAPVSC SGPPPPPPPP VPPPPTGATP PP Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.
Ena/VASP proteins are actin-associated proteins involved in a range of processes dependent on cytoskeleton remodeling and cell polarity such as axon guidance and lamellipodial and filopodial dynamics in migrating cells. EVL enhances actin nucleation and polymerization.Specifications
EVL | |
Polyclonal | |
Unconjugated | |
EVL | |
AI528774; ena/vasodilator-stimulated phosphoprotein-like; Ena/VASP-like protein; Enah/Vasp-like; Ena-vasodilator stimulated phosphoprotein; Evl; Rnb6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51466 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UI08 | |
EVL | |
Recombinant protein corresponding to Human EVL | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only