Learn More
Lenz Laborglasintrumente Evaporating Flasks with Conical Ground Joint, with PUR Coating
Product Code. p-7222615
Click to view available options
Volume (Metric):
50 mL
100 mL
250 mL
2 L
3 L
Unit Size:
Each
Product Code. 11903391
Brand: Lenz Laborglasintrumente 03044037
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Especially for rotary evaporators
  • With center neck and conical joint
  • With highly transparent PUR coating
  • Autoclavable
  • Pear-shaped
  • Material: DURAN™
Specifications
Material Duran Glass
Volume (Metric) 100 mL
Diameter (Metric) 60 mm