Learn More
Lenz Laborglasintrumente Evaporating Flasks with Conical Ground Joint, with PUR Coating
Product Code. 11953391
Change view
Click to view available options
Volume (Metric):
50 mL
100 mL
250 mL
2 L
3 L
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11953391 3 L Each
11903391 100 mL Each
15695585 250 mL Each
11943391 2 L Each
11993381 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11953391 Supplier Lenz Laborglasintrumente Supplier No. 03044076
  • ONLINE EXCLUSIVE PRICE
    259.00€
    182.07€ / Pack of 1
    Save 76.93€
    29% Off
Estimated Shipment: 27-10-2026
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Especially for rotary evaporators
  • With center neck and conical joint
  • With highly transparent PUR coating
  • Autoclavable
  • Pear-shaped
  • Material: DURAN™
Spezifikation
Material Duran Glass
Volume (Metric) 3 L
Diameter (Metric) 175 mm