Learn More
Lenz Laborglasinstrumente™ Evaporating Flask with Conical Ground Joint
Product Code. 11816204
Change view
Click to view available options
Capacity (Metric):
500 mL
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11816204 500 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11816204 Supplier Lenz Laborglasinstrumente™ Supplier No. 03042958
  • ONLINE EXCLUSIVE PRICE
    570.00€
    334.66€ / Pack of 10
    Save 235.34€
    41% Off
Estimated Shipment: 07-12-2026
Especially for use in rotary evaporators. Lenz Laborglasinstrumente™ Evaporating Flasks with Conical Ground Joints can be used to make solutions or to hold, contain, collect or measure chemicals and are manufactured from durable DURAN™ glass.
  • With center neck and conical joint
  • Pear-shaped with inscription label
  • Available in a range of capacities
Specifications
Capacity (Metric) 500 mL
Material Borosilicate Glass, Duran Glass
For Use With (Equipment) Rotary Evaporators
Neck Style Center