Learn More
Heidolph™ Evaporating Flask
Product Code. 10251292 Shop All Heidolph Instruments Products
Change view
Click to view available options
Volume (Metric):
50 mL
2000 mL
3000 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10251292 50 mL Each
10545872 2000 mL Each
10629922 3000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10251292 Supplier Heidolph™ Supplier No. 5147000000
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
For use with rotary evaporators
Evaporating flasks come with a standard NS 29/32 joint size but are also available in NS 24/29
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Glass
Type Evaporating Flask
For Use With (Equipment) Rotary evaporators
Volume (Metric) 50 mL