Learn More
RETSCH Evacuable Pressing Tools
Product Code. p-8003835 Shop All Retsch Products
Click to view available options
Diameter (Metric):
32 mm dia. Pellets
40 mm dia. Pellets
Unit Size:
Each
Product Code. 11944752
Brand: RETSCH 22.458.0017
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Prepare solid samples for XRF analysis. Retsch™ Evacuable Pressing Tools are designed for use with Retsch PP 25 Pellet Presses.
Retsch PP 25 Pellet Press is a compact benchtop unit with particularly simple and safe operation.
  • Produces high quality, stable pellets
  • Easy and safe operation
  • Several pressing tools available
  • Can be evacuated
  • Compact benchtop unit
Specifications
Type Evacuable Pressing Tools
Diameter (Metric) 40 mm dia. Pellets
For Use With (Equipment) Pellet Press PP 25
Weight (Metric) 0.01 kg