Learn More
RETSCH Evacuable Pressing Tools
Product Code. 11954572 Shop All Retsch Products
Change view
Click to view available options
Diameter (Metric):
32 mm dia. Pellets
40 mm dia. Pellets
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
11954572 32 mm dia. Pellets Each
11944752 40 mm dia. Pellets Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11954572 Supplier RETSCH Supplier No. 22.458.0016
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Prepare solid samples for XRF analysis. Retsch™ Evacuable Pressing Tools are designed for use with Retsch PP 25 Pellet Presses.
Retsch PP 25 Pellet Press is a compact benchtop unit with particularly simple and safe operation.
  • Produces high quality, stable pellets
  • Easy and safe operation
  • Several pressing tools available
  • Can be evacuated
  • Compact benchtop unit
Specifications
Type Evacuable Pressing Tools
Diameter (Metric) 32 mm dia. Pellets
For Use With (Equipment) Pellet Press PP 25
Weight (Metric) 1 kg