Learn More
Fisherbrand™ EVA Foam Ice Pans and Buckets with Lids
Product Code. 16544251 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
1 L
2.5 L
4 L
9 L
Color:
Blue
Green
Purple
Includes:
Bucket with Lid
Pan with Lid
Unit Size:
Each
Product Code. 16544251
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Includes
removable lid.
Fisherbrand™EVA Foam Ice Pans and Buckets with Lids provide excellent insulation for any benchtop cryogenic application, without the mess!
  • Sweat-proof ethylene vinyl acetate will not leak
  • Compatible with ice, dry ice, liquid nitrogen, alcohol, and saline solutions
  • Bases and lids feature glove-friendly handles
  • Cleanup is quick and easy with molded pour spouts
Specifications
For Use With (Equipment) Ice, dry ice, liquid nitrogen, alcohol, and saline solutions
Includes Bucket with Lid
Description Bucket with Lid
Material EVA
Type Cryo-Safe Cold Box
Quantity 1
Color Purple
Capacity (Metric) 2.5 L
Autoclavable Not Autoclavable
Temperature Control No