Specifications
| Product Type | EV isolation, exosome isolation, extracellular vesicle isolation |
| Quantity | 25 mL |
| Sufficient For | A 25mL solution is equalivant to 25 reactions (cell culture media) |
| Content And Storage | Store at room temperature. |
| Sample Type | Cell culture supernatant |