Learn More
Thermo Scientific™ Eutech™ Ion Selective Combination Electrode, Epoxy-Body
Description
Eutech™ offers a wide selection of Ion Selective Electrodes (ISE) and meters for the measurement of ammonia, bromide, cadmium, calcium, carbon dioxide, carbonate, chloride, copper, cyanide, fluoride, iodide, lead, nitrate, potassium, silver, sulfide and sodium.ISE responds to ionic activity or concentration in a liquid because of the characteristics of the electrode’s sensing membrane. The ISE develops an electrical potential which is proportional to the activity or concentration of the ion for which the membrane is selective.
Features:
- Epoxy-body
- For Use with: IS meters
Specifications
Specifications
| Material | Epoxy |
| Electrode | Calcium |
| For Use With (Equipment) | Eutech Ion Meters |
| Type | Combination ISE |