Learn More
IKA™ EUROSTAR 60 Control Overhead Stirrer
GREENER_CHOICE
Product Code. p-7163874 Shop All IKA Products
Click to view available options
Plug Type:
EU
Unit Size:
Each
Product Code. 13480567
Brand: IKA™ 0004440000
Includes
IKA, EUROSTAR, 60, control, pverhead, stirrer
Requires
IKA, EUROSTAR, 60, control, pverhead, stirrer
Universal laboratory stirrer designed with a removable wireless controller and a digital TFT display.
  • Comes equipped with a RS 232 and a USB interface to control and document all parameters
  • An integrated torque trend display is provided for the measurement of viscosity changes
  • Safety circuits installed ensures automatic cut-off in an anti-stall or overload conditions
  • Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically
  • Guarantees a constant speed even with changes in viscosities of the sample
  • Multilingual TFT display
  • Programmable functions
  • Integrated temperature measurement
  • Interval operation
  • Timer function
  • Adjustable safety circuit
  • Locked function
  • Infinitely adjustable speed
  • Push-through agitator shafts
  • Overload protection
  • Short-term overload operation
  • Slim casing
  • Quiet operation
  • Error code display
  • H 67.60 temperature sensor and WH 11 WiCo holder included in delivery
Order Info
IKA, EUROSTAR, 60, control, pverhead, stirrer
Certifications
IKA, EUROSTAR, 60, control, pverhead, stirrer
Compliance
IKA, EUROSTAR, 60, control, pverhead, stirrer
Warranty
IKA, EUROSTAR, 60, control, pverhead, stirrer
IKA, EUROSTAR, 60, control, pverhead, stirrer
TRUSTED_SUSTAINABILITY
Specifications
Speed 30 to 2000 rpm
Capacity 40 L
Display TFT
Voltage 230 V
Wattage 176 w
Height (Metric) 267 mm
Width (Metric) 86 mm
Electrical Requirements 230V, 50/60Hz, 176w
Frequency 50/60 Hz
Plug Type EU
Chuck Size 0.5 to 10 mm
Includes RS 232 and a USB interface
Material Alu-Cast Coating/Thermoplastic Polymer
Model EUROSTAR 60 Control
Motor Type Brushless DC
Temperature Range (Metric) -10°C to +350°C
Torque 60 Ncm
Viscosity 50,000 mPas