Learn More
IKA™ EUROSTAR 100 Overhead Stirrers
GREENER_CHOICE
Product Code. 13490567 Shop All IKA Products
Click to view available options
Model:
EUROSTAR 100 Control
EUROSTAR 100 Digital
Dimensions (L x W x H):
H x W x dia.: 248 x 86 x 208 mm
H x W x dia.: 267 x 86 x 230 mm
Unit Size:
Each
Product Code. 13490567
Brand: IKA™ 0004238100
Only null left
Add to basket
Add to basket
Use this powerful laboratory mixer for a variety of applications. IKA™ EUROSTAR 100 Overhead Stirrer automatically adjusts the speed through microprocessor controlled technology.
  • Equipped with a RS 232 and a USB interface to control and document all parameters
  • Integrated torque trend display is provided for the measurement of viscosity changes
  • Safety circuits installed ensures automatic cut-off in an anti-stall or overload conditions
  • Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically
  • Guarantees constant speed even with changes in viscosities of the sample
  • Multilingual TFT display
  • Programmable functions
  • Integrated temperature measurement
  • Interval operation
  • Reverse function
  • Timer function
  • Adjustable safety circuit
  • Locked function
  • Infinitely adjustable speed
  • Push-through agitator shafts
  • Overload protection
  • Short-term overload operation
  • Quiet operation
  • Error code Display
  • H 67.60 temperature sensor and WH 11 WiCo holder included in delivery
TRUSTED_SUSTAINABILITY
Specifications
Speed 30 to 1300 rpm
Capacity 100 L
Display 7 Segment LED
Voltage 230 V
Wattage 186 w
Dimensions (L x W x H) H x W x dia.: 248 x 86 x 208 mm
Height (Metric) 248 mm
Width (Metric) 86 mm
Electrical Requirements 230V, 50/60Hz, 186w
Frequency 50/60 Hz
Material Alu-Cast Coating/Thermoplastic Polymer
Model EUROSTAR 100 Digital
Temperature Range (Metric) 5°C to 40°C
Torque 100 Ncm
Viscosity 70,000 mPas