Learn More
Euro Protection™ Eurmask Uno
Click to view available options
:
COMPATIBILITY: 22130, 22140, 22150, 22160, 22170, 22210
Description
- Half mask for 1 threaded cartridge
- Body: TPE thermoplastic rubber
- Harness: Fabric and rubber elastic strap
- Valves: Silicone
- Adjustable straps
Specifications
Specifications
| Certifications/Compliance | EN 140 :1998 |
| Material | TPE Thermoplastic rubber, silicone |
| For Use With (Equipment) | COMPATIBILITY: 22130, 22140, 22150, 22160, 22170, 22210 |
| Quantity | 1/Pk., 16Pk./Cs. |
| Weight (Metric) | 115 g |