Learn More
SCHOTT AG Lighting and Imaging™ EU Power Cord
Product Code. 10798630 Shop All Schott Products
Click to view available options
Type:
EU Power Cord
Unit Size:
Each
Product Code. 10798630
Brand: SCHOTT AG Lighting and Imaging™ 400051
Only null left
Add to basket
Edge
Add to basket
Power Cable with EU plug.
  • Power cord with EU plug.
  • Cable length: 1.8 m
  • For use with KL1600LED, KL2500LED, KL1500HAL and all VisiLED Controllers.
Specifications
Type EU Power Cord
For Use With (Equipment) KL 1600 LED, KL 2500 LED, KL 1500 HAL, VisiLED Controllers
Length (English) 5.9 ft.
Length (Metric) 1.8 m