Learn More
Cytiva Ettan IPGPhor™ Strip Holder
Product Code. 10127004 Shop All Cytiva Products
Change view
Click to view available options
Size: 24cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10127004 Size: 24cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10127004 Supplier Cytiva Supplier No. 80646988
Estimated Shipment: 03-11-2026
Specifications
Type Strip holder
Includes 6 x Strip holder cover 1 x 60 ml Strip holder cleaning solution
Length (Metric) 24 cm