Learn More
Cytiva Ettan™ DALT Staining Tray Set
Product Code. 10627875 Shop All Cytiva Products
Click to view available options
For Use With (Equipment):
Ettan DALTtwelve Large Vertical System and Ettan DALTsix Large Vertical System
Unit Size:
1 set
Product Code. 10627875
Brand: Cytiva 80646817
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For Use With: Ettan DALTtwelve Large Vertical System and Ettan DALTsix Large vertical System
After the second-dimension run is complete, the large-format gels can be stained using DALT staining tray set and PlusOne silver staining kit, Protein. This combination ensures reliable detection with high sensitivity.The set includes two stainless steel trays for staining solutions; one perforated stainless steel tray that seats within the staining trays; and a transparent plastic cover. The perforated insert supports and restrains gels for transfer between staining trays while allowing staining solutions to drain rapidly.
  • For Use With: Ettan DALTtwelve Large Vertical System and Ettan DALTsix Large vertical System
Kit Contains:
  • 2 x Tray for staining solutions (stainless steel)
  • 1 x Perforated tray that seats within the staining trays (23x42 cm, stainless steel)
  • 1 x Cover (transparent plastic)
  • Instructions
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Reproducible high resolution, high throughput 2D electrophoresis
For Use With (Equipment) Ettan DALTtwelve Large Vertical System and Ettan DALTsix Large Vertical System
Type Staining Tray Set
Includes 2 Tray for staining solutions, 1 Perforated tray that seats within the staining trays, 1 Plastic cover, instructions
Quantity 1 Kit