Learn More
Etofenamate, TRC
Product Code. 30135438
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135438 1 g 1g
30135421 100 mg 100mg
30005461 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135438 Supplier Toronto Research Chemicals Supplier No. TRCE9331351G
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 30544-47-9
Molecular Formula C18 H18 F3 N O4
Molecular Weight (g/mol) 369.34
Synonym Etofenamate, 2-(2-Hydroxyethoxy)ethyl 2-[[3-(trifluoromethyl)phenyl]amino]benzoate, Benzoic acid, 2-[[3-(trifluoromethyl)phenyl]amino]-, 2-(2-hydroxyethoxy)ethyl ester, Anthranilic acid, N-(α,α,α-trifluoro-m-tolyl)-, 2-(2-hydroxyethoxy)ethyl ester (8CI), 2-(2-Hydroxyethoxy)ethyl flufenamate, 2-[[3-(Trifluoromethyl)phenyl]amino]benzoic acid 2-(2-hydroxyethoxy)ethyl ester, B 577, Bay-d 1107, Bayrogel, Etofenamate, Glasel, Reumon, Rheumon, Rheumon gel, TV 485, Traumon Gel, WHR 5020
IUPAC Name 2-(2-hydroxyethoxy)ethyl 2-[3-(trifluoromethyl)anilino]benzoate
SMILES OCCOCCOC(=O)c1ccccc1Nc2cccc(c2)C(F)(F)F
Specifications
Quantity 1 g
InChI Formula InChI=1S/C18H18F3NO4/c19-18(20,21)13-4-3-5-14(12-13)22-16-7-2-1-6-15(16)17(24)26-11-10-25-9-8-23/h1-7,12,22-23H,8-11H2
Formula Weight 369.1188
Percent Purity >95%
Chemical Name or Material Etofenamate
Grade HPLC