missing translation for 'onlineSavingsMsg'
Learn More

Ethynyl Estradiol, TRC

Product Code. 30136896
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
1g
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136896 1 g 1g
18091840 2.5 g 2.5g
18002640 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136896 Supplier Toronto Research Chemicals Supplier No. TRCE6851001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-63-6
Molecular Formula C20 H24 O2
Molecular Weight (g/mol) 296.4
Synonym (17alpha)-19-Norpregna-1,3,5(10)-trien-20-yne-3,17-diol, 19-Nor-17alpha-pregna-1,3,5(10)-trien-20-yne-3,17-diol, 17-Ethinyl-3,17-estradiol, 17-Ethinylestradiol, 17-Ethynyl-3,17-dihydroxy-1,3,5-oestratriene, 17-Ethynylestra-1,3,5(10)-triene-3,17beta-diol, 17-Ethynylestradiol, 17-Nor-17alpha-pregna-1,3,5-(10)-trien-20-yne-3,17-diol, 17alpha-Ethinyl-1,3,5(10)-estratriene-3,17-diol, 17alpha-Ethinyl-17beta-estradiol, 17alpha-Ethinyl-3,17-dihydroxy-delta1,3,5-estratriene, 17alpha-Ethinylestra-1,3,5(10)-triene-3,17beta-diol, 17alpha-Ethinylestradiol, 17alpha-Ethynylestra-1,3,5(10)-triene-3,17beta-diol, 17alpha-Ethynylestradiol, 19-Nor-17alpha-pregna-1,3,5(10)-trien-20-yne-3,17beta-diol, Amenoron, Chee-O-Gen, Chee-O-Genf, Diogyn E, Dyloform, Esteed, Estigyn, Estinyl, Eston-E, Estoral, Estorals, 17-Ethynylestradiol, Ethidol, Ethinoral, Ethinylestradiol, Ethinyloestradiol, Ethynylestradiol, Ethynyloestradiol, Eticyclin, Eticyclol, Etinestrol, Etinestryl, Etinoestryl, Etistradiol, Follicoral, Ginestrene, Inestra, Linoral, Lynoral, Menolyn, Microfollin, NSC 10973, Novestrol, Oradiol, Orestralyn, Palonyl, Perovex, Primogyn, Primogyn C, Primogyn M, Progynon C, Prosexol, Spanestrin, neo-Estrone
IUPAC Name (8R,9S,13S,14S,17R)-17-ethynyl-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthrene-3,17-diol
SMILES C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1CC[C@@]2(O)C#C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C20H24O2/c1-3-20(22)11-9-18-17-6-4-13-12-14(21)5-7-15(13)16(17)8-10-19(18,20)2/h1,5,7,12,16-18,21-22H,4,6,8-11H2,2H3/t16-,17-,18+,19+,20+/m1/s1
Formula Weight 296.1776
Percent Purity >95%
Chemical Name or Material Ethinylestradiol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.