Learn More
Ethylenediaminetetraacetic Acid, Tetrasodium Salt Tetrahydrate, Extra Pure, SLR, Fisher Chemical
Product Code. 10070100 Shop All Fisher Chemical Products
Click to view available options
:
250g, Plastic powder jar
Product Code. 10070100
Brand: Fisher Chemical D/0750/50
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Melting Point 300°C
CAS 13235-36-4
Color White
pH 11 to 12
Physical Form Solid
Quantity 250 g
Molecular Formula C10H20N2Na4O12
MDL Number 150027
InChI Key XFLNVMPCPRLYBE-UHFFFAOYSA-J
SMILES O.O.O.O.[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
IUPAC Name tetrasodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate tetrahydrate
Molecular Weight (g/mol) 452.23
Formula Weight 452.23g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethylenediaminetetraacetic acid, tetrasodium salt tetrahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Ethylenediaminetetraacetate, tetrasodiumtetrahydrate, Tetrasodium EDTA