missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid, (EDTA), 0.5M Solution, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals

Catalog Number p-7082789
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Nalgene bottle
This item is not returnable. View return policy
6381-92-6
C10H18N2Na2O10
372.238
MFCD00150037
OVBJJZOQPCKUOR-UHFFFAOYSA-L
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
44120005
disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[Na+].[Na+]
This item is not returnable. View return policy
6381-92-6
C10H18N2Na2O10
372.238
MFCD00150037
OVBJJZOQPCKUOR-UHFFFAOYSA-L
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
44120005
disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[Na+].[Na+]

Chelating agent used in many enzyme buffers and at higher concentrations, as enzyme inactivator

Chelating agent used in many enzyme buffers and, at higher concentrations, as an enzyme inactivator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Packaging Nalgene bottle
Quantity 100 mL
Beilstein 3900609
Grade Molecular Biology, Ultrapure
Concentration or Composition (by Analyte or Components) 0.5 M
Physical Form Solution
Chemical Name or Material Ethylenediaminetetraacetic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'