Learn More
Ethylenediamine Dihydrochloride, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Ethylenediamine Dihydrochloride |
| CAS | 333-18-6 |
| Molecular Formula | C2H8N2 . 2(HCl) |
| Quantity | 10 g |
| Synonym | Ethylenediamine dihydrochloride, 1,2-Ethanediamine, hydrochloride (1:2), 1,2-Ethanediamine, dihydrochloride (9CI), Ethylenediamine, dihydrochloride (8CI), 1,2-Diaminoethane dihydrochloride, Chlor-ethamine, Dimethylenediamine dihydrochloride, Ethylenediamine hydrochloride, Ethylenediamine hydrochloride (1:2), Ethylenediammonium dichloride, 1,2-Ethylenediamine dihydrochloride, Ethane-1,2-diamine dihydrochloride |
| InChI Formula | InChI=1S/C2H8N2/c3-1-2-4/h1-4H2 |
| SMILES | Cl.Cl.NCCN |
| Molecular Weight (g/mol) | 133.02 |
| Formula Weight | 132.0221 |