Learn More
Ethylene-d4-diamine, CDN
Specifications
Specifications
| Chemical Name or Material | Ethylene-d4 Diamine |
| CAS | 37164-19-5 |
| Molecular Formula | C2 D4 H4 N2 |
| Quantity | 0.5 g |
| Synonym | 1,2-Ethanediamine-d4, 1,2-Diaminoethane-d4, Chlor-ethamine-d4, Dimethylenediamine-d4, 1,2-Ethane-1,1,2,2-d4-diamine (9 CI), 1,2-Ethane-1,1,2,2-d4-diamine (9 CI), 1,2-Ethane-1,1,2,2-d4-diamine (9 CI), 1,2-Ethane-1,1,2,2-d4-diamine (9 CI), 1,2-Ethane-1,1,2,2-d4-diamine (9 CI) |
| InChI Formula | InChI=1 S/C2H8N2/c3-1-2-4/h1-4H2/i1D2,2D2 |
| SMILES | [2 H]C([2 H])(N)C([2 H])([2 H])N |
| IUPAC Name | 1,1,2,2-tetradeuterioethane-1,2-diamine |
| Molecular Weight (g/mol) | 64.12 |
| Formula Weight | 64.0939 g/mol |
| Show More |