Learn More
Ethyl [(tert-Butyldimethylsilyl)oxy]acetate, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl [(tert-Butyldimethylsilyl)oxy]acetate |
| CAS | 67226-78-2 |
| Molecular Formula | C10 H22 O3 Si |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C10H22O3Si/c1-7-12-9(11)8-13-14(5,6)10(2,3)4/h7-8H2,1-6H3 |
| SMILES | CCOC(=O)CO[Si](C)(C)C(C)(C)C |
| IUPAC Name | ethyl 2-[tert-butyl(dimethyl)silyl]oxyacetate |
| Molecular Weight (g/mol) | 218.37 |
| Formula Weight | 218.1338 |