Learn More
Ethyl Pyruvate-3,3,3-d3, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 2-Oxopropanoic-3,3,3-d3 acid ethyl ester |
| CAS | 66966-38-9 |
| Molecular Formula | C5 2H3 H5 O3 |
| Quantity | 0.05 g |
| Synonym | Propanoic-3,3,3-d3 acid, 2-oxo-, ethyl ester (9 CI), 2-Oxopropanoic-3,3,3-D3 acid ethyl ester, Pyruvic-3,3,3-d3 acid ethyl ester, Ethyl pyruvate-3,3,3-d3, Ethyl pyruvate-d3 |
| InChI Formula | InChI=1 S/C5H8O3/c1-3-8-5(7)4(2)6/h3H2,1-2H3/i2D3 |
| SMILES | [2 H]C([2 H])([2 H])C(=O)C(=O)OCC |
| IUPAC Name | ethyl 3,3,3-trideuterio-2-oxopropanoate |
| Molecular Weight (g/mol) | 119.13 |
| Formula Weight | 119.0662 g/mol |
| Show More |