Learn More
Ethyl oxalyl chloride, 98%
Product Code. p-7022057
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. 11437616
Brand: Thermo Scientific Alfa Aesar A14653.14
Request Bulk or Custom Format
CAS: 4755-77-5 | C4H5ClO3 | 136.531 g/mol
Ethyl oxalyl chloride is used in the synthesis of 2-oxo-3-alkenoic esters and alpha-keto ester in good yield. It acts as a reactant in the preparation of oxyoxalamide derivatives as an epoxide hydrolase inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl oxalyl chloride is used in the synthesis of 2-oxo-3-alkenoic esters and alpha-keto ester in good yield. It acts as a reactant in the preparation of oxyoxalamide derivatives as an epoxide hydrolase inhibitor.
Solubility
Slightly miscible with water.
Notes
Moisture sensitive. Incompatible with bases, oxidizing agents and alcohols.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4755-77-5
Molecular Formula C4H5ClO3
Molecular Weight (g/mol) 136.531
MDL Number MFCD00000706
InChI Key OWZFULPEVHKEKS-UHFFFAOYSA-N
Synonym ethyl chlorooxoacetate, ethyl oxalyl chloride, ethyl chloroglyoxylate, ethoxalyl chloride, ethyl oxalyl monochloride, acetic acid, chlorooxo-, ethyl ester, oxalic acid monoethyl ester chloride, monoethyl oxaloyl chloride, oxalyl chloride, ethyl ester, ethyl chlorocarbonyl formate
PubChem CID 20884
IUPAC Name ethyl 2-chloro-2-oxoacetate
SMILES CCOC(=O)C(=O)Cl
Specifications
Density 1.226
Boiling Point 134°C to 135°C
Flash Point 41°C (105°F)
Linear Formula ClCOCO2CH2CH3
Refractive Index 1.416
Quantity 25 g
UN Number UN2920
Beilstein 506725
Sensitivity Moisture sensitive
Solubility Information Slightly miscible with water.
Formula Weight 136.53
Percent Purity 98%
Chemical Name or Material Ethyl oxalyl chloride