Learn More
Ethyl Nicotinate-2,4,5,6-d4, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Ethyl Nicotinate-d4 |
| CAS | 66148-16-1 |
| Molecular Formula | C8 2H4 H5 N O2 |
| Quantity | 0.5 g |
| InChI Formula | InChI=1 S/C8H9NO2/c1-2-11-8(10)7-4-3-5-9-6-7/h3-6 H,2H2,1H3/i3D,4 D,5 D,6 D |
| SMILES | [2 H]c1nc([2 H])c(C(=O)OCC)c([2 H])c1[2 H] |
| IUPAC Name | ethyl 2,4,5,6-tetradeuteriopyridine-3-carboxylate |
| Molecular Weight (g/mol) | 155.19 |
| Formula Weight | 155.0884 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |