Learn More
Ethyl isocyanate, 98%
Product Code. p-7232713
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. 15473277
Brand: Thermo Scientific Alfa Aesar B25180.06
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Boiling Point 60°C to 61°C
Quantity 5 g
Percent Purity 98%
Physical Form Liquid