missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl-d5-benzene, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Ethylbenzene (ethyl-D5) |
| CAS | 38729-11-2 |
| Molecular Formula | C8 2H5 H5 |
| Quantity | 0.5 g |
| Synonym | Benzene, ethyl-1,1,2,2,2-d5-, Benzene, ethyl-d5- (9 CI), Ethyl-1,1,2,2,2-d5benzene, Ethylbenzene (ethyl-D5) |
| InChI Formula | InChI=1 S/C8H10/c1-2-8-6-4-3-5-7-8/h3-7 H,2H2,1H3/i1D3,2D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])c1ccccc1 |
| IUPAC Name | 1,1,2,2,2-pentadeuterioethylbenzene |
| Molecular Weight (g/mol) | 111.2 |
| Formula Weight | 111.1096 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction