Learn More
Ethyl 5-benzyloxyindole-2-carboxylate, 97%
Product Code. 11321587
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11321587 5 g 5g
11311587 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11321587 Supplier Thermo Scientific Alfa Aesar Supplier No. L04440.06
Request Bulk or Custom Format
CAS: 37033-95-7 | C18H17NO3 | 295.338 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 37033-95-7
Molecular Formula C18H17NO3
Molecular Weight (g/mol) 295.338
MDL Number MFCD00022702
InChI Key DCIFXYFKVKDOLL-UHFFFAOYSA-N
Synonym ethyl 5-benzyloxy-1h-indole-2-carboxylate, ethyl 5-benzyloxyindole-2-carboxylate, 5-benzyloxyindole-2-carboxylic acid ethyl ester, 5-benzyloxy-2-carbethoxyindole, ethyl 5-benzyloxy indole-2-carboxylate, 5-benzyloxyindole-2-carboxylic acid ethylester, ethyl 5-phenylmethoxy-1h-indole-2-carboxylate, 5-phenylmethoxy-1h-indole-2-carboxylic acid ethyl ester, 5-benzyloxyindole-2-carboxylic acid*ethyl ester c, 1h-indole-2-carboxylic acid, 5-phenylmethoxy-, ethyl ester
PubChem CID 95721
IUPAC Name ethyl 5-phenylmethoxy-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=C(N1)C=CC(=C2)OCC3=CC=CC=C3
Specifications
Melting Point 160°C to 165°C
Quantity 5 g
Beilstein 281129
Formula Weight 295.34
Percent Purity 97%
Chemical Name or Material Ethyl 5-benzyloxyindole-2-carboxylate