Learn More
Ethyl 4-Quinazolone-2-carboxylate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 29113-33-5 |
| Molecular Formula | C11H10N2O3 |
| MDL Number | MFCD00051583 |
| Quantity | 100 g |
| SMILES | O=C(C(N1)=NC2=C(C=CC=C2)C1=O)OCC |
| Molecular Weight (g/mol) | 218.21 |
| Percent Purity | 98% |
| Chemical Name or Material | Ethyl 4-Quinazolone-2-carboxylate |