missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Hydrazinylbenzoate, TRC

Product Code. 30372727
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30372727 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30372727 Supplier Toronto Research Chemicals Supplier No. TRCE938768250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 4-Hydrazinylbenzoate
CAS 14685-90-6
Molecular Formula C9 H12 N2 O2
Quantity 250 mg
Synonym 4-(Ethoxycarbonyl)phenylhydrazine, 4-(Hydrazinyl)benzoic Acid Ethyl Ester, 4-Carbethoxyphenylhydrazine, Ethyl p-hydrazinobenzoate
InChI Formula InChI=1S/C9H12N2O2/c1-2-13-9(12)7-3-5-8(11-10)6-4-7/h3-6,11H,2,10H2,1H3
SMILES CCOC(=O)c1ccc(NN)cc1
IUPAC Name ethyl 4-hydrazinylbenzoate
Molecular Weight (g/mol) 180.204
Formula Weight 180.09
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.