Learn More
Ethyl 4-aminophenylacetate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 5438-70-0 |
| Molecular Formula | C10H13NO2 |
| MDL Number | MFCD00017569 |
| Quantity | 25 g |
| SMILES | O=C(OCC)CC1=CC=C(N)C=C1 |
| Molecular Weight (g/mol) | 179.22 |
| Percent Purity | 98% |
| Chemical Name or Material | Ethyl 4-aminophenylacetate |