missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-aminobenzoate, 98%

Product Code. 11485073
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
This item is not returnable. View return policy

Product Code. 11485073

Brand: Thermo Scientific Alfa Aesar A12754.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 94-09-7 | C9H11NO2 | 165.19 g/mol

Ethyl 4-aminobenzoate is used as an anesthetic and utilized for topical pain reliever. It is mixed with antipyrine to get A/B otic drops, which is used in the removal of earwax as well as relieve ear from pain. It exhibits analgesic and antipruritic effect mainly used for wounds, ulcers, mucosal surfaces and hemorrhoids narcotic pain. Further, it is a main constituent of anesthetic ointments. In addition to this, it is effective to toothache, sore throat, oral ulcers, hemorrhoids and anal fissure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-aminobenzoate is used as an anesthetic and utilized for topical pain reliever. It is mixed with antipyrine to get A/B otic drops, which is used in the removal of earwax as well as relieve ear from pain. It exhibits analgesic and antipruritic effect mainly used for wounds, ulcers, mucosal surfaces and hemorrhoids narcotic pain. Further, it is a main constituent of anesthetic ointments. In addition to this, it is effective to toothache, sore throat, oral ulcers, hemorrhoids and anal fissure.

Solubility
Soluble in ethanol, chloroform, ethyl ether and dilute acids. Sparingly soluble in water

Notes
Incompatible with strong oxidizing agents, reducing agents, acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-09-7
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00007892
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
Synonym benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
PubChem CID 2337
ChEBI CHEBI:116735
IUPAC Name ethyl 4-aminobenzoate
SMILES CCOC(=O)C1=CC=C(N)C=C1

Specifications

Melting Point 88°C to 92°C
Boiling Point 310°C
Flash Point >110°C (230°F)
Odor Odorless
Quantity 1000 g
Beilstein 638434
Merck Index 14,1086
Solubility Information Soluble in ethanol,chloroform,ethyl ether and dilute acids. Sparingly soluble in water
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material Ethyl 4-aminobenzoate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 4-aminobenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Ethyl 4-aminobenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.