Learn More
Ethyl 3-[(Tetrahydropyranyl)oxy]butanoate, TRC
Product Code. 30376378
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30376378 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30376378 Supplier Toronto Research Chemicals Supplier No. TRCE926483250MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ethyl 3-[(Tetrahydropyranyl)oxy]butanoate
CAS 104372-23-8
Molecular Formula C5 H5 N O3 S
Quantity 250 mg
Synonym 3-[(Tetrahydro-2H-pyran-2-yl)oxy]-butanoic Acid Ethyl Ester, Ethyl 3-((Tetrahydro-2H-pyran-2-yl)oxy)butanoate
InChI Formula InChI=1S/C5H5NO3S/c7-10(8,9)5-2-1-3-6-4-5/h1-4H,(H,7,8,9)
SMILES OS(=O)(=O)c1cccnc1
IUPAC Name pyridine-3-sulfonic acid
Molecular Weight (g/mol) 159.163
Formula Weight 158.999