Learn More
Ethyl 3-(pyridin-2-ylamino)propanoate, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 103041-38-9 |
| Molecular Formula | C10H14N2O2 |
| MDL Number | MFCD09833621 |
| Quantity | 100 g |
| SMILES | C(C)OC(CCNC1=NC=CC=C1)=O |
| Molecular Weight (g/mol) | 194.23 |
| Percent Purity | 95% |
| Chemical Name or Material | Ethyl 3-(pyridin-2-ylamino)propanoate |